The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017514.4 | NP_059984.3 | 24 - 25 | 4469 | 67 |
XM_005274705.4 | XP_005274762.2 | 24 - 25 | 4409 | 67 | |
XM_005274706.4 | XP_005274763.2 | 24 - 25 | 4141 | 67 | |
XM_006724829.3 | XP_006724892.1 | 24 - 25 | 4373 | 67 | |
XM_011531183.2 | XP_011529485.1 | 23 - 24 | 3833 | 67 | |
GenBank mRNA | X87852.1 | - | 24 - 25 | 4478 | 67 |