The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032737.3 | NP_116126.3 | 5 - 6 | 897 | 59 |
GenBank mRNA | BC006551.2 | - | 5 - 6 | 862 | 59 |
BT007441.1 | - | 5 - 6 | 802 | 59 | |
KP137589.1 | - | 2 - 3 | 451 | 59 | |
M94362.1 | - | 4 - 5 | 550 | 59 |