The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261826.1 | NP_001248755.1 | 13 - 14 | 1476 | 106 |
NM_003938.6 | NP_003929.4 | 13 - 14 | 1476 | 106 | |
XM_006722932.2 | XP_006722995.1 | 13 - 14 | 1539 | 106 | |
XM_017027422.1 | XP_016882911.1 | 8 - 9 | 773 | 106 | |
GenBank mRNA | AB208804.1 | - | 13 - 14 | 1460 | 106 |
AF002163.1 | - | - | 1564 | 106 | |
AF130042.1 | - | 11 - 12 | 1035 | 106 | |
BC005142.2 | - | 13 - 14 | 1304 | 106 | |
BC010065.2 | - | 13 - 14 | 1455 | 106 | |
U91930.1 | - | 13 - 14 | 1189 | 106 |