The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261826.1 | NP_001248755.1 | 25 - 26 | 3161 | 79 |
NM_003938.6 | NP_003929.4 | 23 - 24 | 2975 | 79 | |
XM_006722932.2 | XP_006722995.1 | 25 - 26 | 3188 | 79 | |
XM_017027422.1 | XP_016882911.1 | 20 - 21 | 2458 | 79 | |
GenBank mRNA | AB208804.1 | - | 25 - 26 | 3145 | 79 |
AF002163.1 | - | - | 3063 | 79 | |
U91930.1 | - | 25 - 26 | 2838 | 79 |