The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160213.1 | NP_001153685.1 | 19 - 20 | 3048 | 59 |
NM_031293.2 | NP_112583.2 | 18 - 19 | 2870 | 59 | |
XM_011523357.2 | XP_011521659.1 | 18 - 19 | 2975 | 59 | |
XM_011523358.2 | XP_011521660.1 | 18 - 19 | 2915 | 59 | |
XM_011523360.2 | XP_011521662.1 | 18 - 19 | 2938 | 59 | |
XM_011523361.2 | XP_011521663.1 | 18 - 19 | 3413 | 59 | |
XM_011523362.2 | XP_011521664.1 | 17 - 18 | 2720 | 59 | |
GenBank mRNA | AF239683.1 | - | 3 - 4 | 331 | 59 |
AK093285.1 | - | 15 - 16 | 2236 | 59 | |
AK122925.1 | - | 19 - 20 | 3048 | 59 | |
AK315004.1 | - | 18 - 19 | 2851 | 59 | |
AL136795.1 | - | 18 - 19 | 2830 | 59 | |
BC028423.2 | - | 18 - 19 | 2872 | 59 |