Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PMFBP1
Entrez Gene ID
83449
Gene Name
polyamine modulated factor 1 binding protein 1
Gene Aliases
-
Chromosome Location
Chr.16: 72116696 - 72173908 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031293.2 NP_112583.2 1 - 2 115 117
XM_011523357.2 XP_011521659.1 1 - 2 205 117
GenBank mRNA AK093285.1 - 1 - 2 111 117
AL136795.1 - 1 - 2 75 117
BC028423.2 - 1 - 2 102 117
DB019901.1 - 1 - 2 205 117

View Full Product Details