Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PMFBP1
Entrez Gene ID
83449
Gene Name
polyamine modulated factor 1 binding protein 1
Gene Aliases
-
Chromosome Location
Chr.16: 72116696 - 72173908 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160213.1 NP_001153685.1 5 - 6 764 87
NM_031293.2 NP_112583.2 4 - 5 586 87
XM_011523357.2 XP_011521659.1 4 - 5 676 87
XM_011523358.2 XP_011521660.1 4 - 5 616 87
XM_011523360.2 XP_011521662.1 4 - 5 639 87
XM_011523361.2 XP_011521663.1 4 - 5 1114 87
XM_011523362.2 XP_011521664.1 3 - 4 421 87
GenBank mRNA AK093285.1 - 4 - 5 582 87
AK122925.1 - 5 - 6 764 87
AK315004.1 - 4 - 5 567 87
AL136795.1 - 4 - 5 546 87
BC028423.2 - 4 - 5 573 87

View Full Product Details