Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PMFBP1
Entrez Gene ID
83449
Gene Name
polyamine modulated factor 1 binding protein 1
Gene Aliases
-
Chromosome Location
Chr.16: 72116696 - 72173908 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160213.1 NP_001153685.1 10 - 11 1555 70
NM_031293.2 NP_112583.2 9 - 10 1377 70
XM_011523357.2 XP_011521659.1 9 - 10 1467 70
XM_011523358.2 XP_011521660.1 9 - 10 1407 70
XM_011523360.2 XP_011521662.1 9 - 10 1430 70
XM_011523361.2 XP_011521663.1 9 - 10 1905 70
XM_011523362.2 XP_011521664.1 8 - 9 1212 70
GenBank mRNA AK093285.1 - 9 - 10 1373 70
AK097465.1 - 5 - 6 614 70
AK122925.1 - 10 - 11 1555 70
AK315004.1 - 9 - 10 1358 70
AL136795.1 - 9 - 10 1337 70
BC028423.2 - 9 - 10 1364 70

View Full Product Details