The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000362.4 | NP_000353.1 | 2 - 3 | 1387 | 114 |
GenBank mRNA | AK314871.1 | - | 2 - 3 | 496 | 114 |
BC014277.2 | - | 2 - 3 | 444 | 114 | |
BT006848.1 | - | 2 - 3 | 201 | 114 | |
CR456593.1 | - | 2 - 3 | 249 | 114 | |
L15078.1 | - | 2 - 3 | 159 | 114 | |
S78453.1 | - | 2 - 3 | 477 | 114 | |
U02571.1 | - | 2 - 3 | 482 | 114 | |
U14394.1 | - | 2 - 3 | 488 | 114 | |
U67195.1 | - | 2 - 3 | 479 | 114 | |
X76227.1 | - | 2 - 3 | 271 | 114 |