Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNX6
Entrez Gene ID
58533
Gene Name
sorting nexin 6
Gene Aliases
MSTP010, TFAF2
Chromosome Location
Chr.14: 34561409 - 34630160 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021249.3 NP_067072.3 4 - 5 541 88
NM_152233.2 NP_689419.2 5 - 6 436 88
XM_006720224.1 XP_006720287.1 4 - 5 323 88
GenBank mRNA AF121856.1 - 5 - 6 427 88
AK292250.1 - 5 - 6 419 88
BC001798.1 - 4 - 5 541 88
BG722853.1 - 5 - 6 462 88
U83194.1 - 5 - 6 404 88

View Full Product Details