Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PALMD
Entrez Gene ID
54873
Gene Name
palmdelphin
Gene Aliases
C1orf11, PALML
Chromosome Location
Chr.1: 99645875 - 99694541 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017734.4 NP_060204.1 3 - 4 688 134
GenBank mRNA AF262379.1 - 3 - 4 619 134
AJ312214.1 - 3 - 4 530 134
AK000278.1 - 3 - 4 450 134
AL390971.1 - 3 - 4 618 134
AL390973.1 - 3 - 4 414 134
BC012570.1 - 3 - 4 503 134
KU178657.1 - 3 - 4 245 134
KU178658.1 - 3 - 4 245 134

View Full Product Details