The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017734.4 | NP_060204.1 | 7 - 8 | 2050 | 66 |
GenBank mRNA | AF262379.1 | - | 7 - 8 | 1981 | 66 |
AJ312214.1 | - | 7 - 8 | 1892 | 66 | |
AK000278.1 | - | 7 - 8 | 1812 | 66 | |
AK024654.1 | - | 2 - 3 | 1113 | 66 | |
AL390969.1 | - | 1 - 2 | 854 | 66 | |
AL390970.1 | - | 1 - 2 | 1079 | 66 | |
AL390972.1 | - | 7 - 8 | 1489 | 66 | |
AL390973.1 | - | 7 - 8 | 1776 | 66 | |
BC012570.1 | - | 7 - 8 | 1865 | 66 | |
BX648213.1 | - | 4 - 5 | 4765 | 66 | |
KU178657.1 | - | 7 - 8 | 1607 | 66 |