The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015104.2 | NP_055919.2 | 12 - 13 | 1818 | 67 |
XM_005273849.3 | XP_005273906.1 | 12 - 13 | 1809 | 67 | |
XM_005273850.3 | XP_005273907.1 | 12 - 13 | 1809 | 67 | |
XM_011544863.2 | XP_011543165.1 | 12 - 13 | 1809 | 67 | |
XM_011544864.2 | XP_011543166.1 | 12 - 13 | 1809 | 67 | |
XM_011544865.2 | XP_011543167.1 | 12 - 13 | 1802 | 67 | |
XM_011544866.2 | XP_011543168.1 | 12 - 13 | 1809 | 67 | |
XM_011544867.2 | XP_011543169.1 | 12 - 13 | 1809 | 67 | |
GenBank mRNA | AB007864.1 | - | 12 - 13 | 1759 | 67 |
AK307684.1 | - | 12 - 13 | 1777 | 67 | |
BC110650.1 | - | 12 - 13 | 1818 | 67 |