The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015104.2 | NP_055919.2 | 15 - 16 | 2317 | 79 |
XM_005273849.3 | XP_005273906.1 | 15 - 16 | 2308 | 79 | |
XM_005273850.3 | XP_005273907.1 | 15 - 16 | 2308 | 79 | |
XM_011544863.2 | XP_011543165.1 | 15 - 16 | 2308 | 79 | |
XM_011544864.2 | XP_011543166.1 | 15 - 16 | 2308 | 79 | |
XM_011544865.2 | XP_011543167.1 | 15 - 16 | 2301 | 79 | |
XM_011544866.2 | XP_011543168.1 | 15 - 16 | 2308 | 79 | |
XM_011544867.2 | XP_011543169.1 | 15 - 16 | 2308 | 79 | |
GenBank mRNA | AB007864.1 | - | 15 - 16 | 2258 | 79 |
BC110650.1 | - | 15 - 16 | 2317 | 79 |