The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015104.2 | NP_055919.2 | 31 - 32 | 4577 | 73 |
XM_005273849.3 | XP_005273906.1 | 31 - 32 | 4550 | 73 | |
XM_005273850.3 | XP_005273907.1 | 31 - 32 | 4544 | 73 | |
XM_011544863.2 | XP_011543165.1 | 31 - 32 | 4574 | 73 | |
XM_011544864.2 | XP_011543166.1 | 31 - 32 | 4559 | 73 | |
XM_011544865.2 | XP_011543167.1 | 31 - 32 | 4567 | 73 | |
XM_011544866.2 | XP_011543168.1 | 31 - 32 | 4574 | 73 | |
XM_011544867.2 | XP_011543169.1 | 29 - 30 | 4150 | 73 | |
XM_011544868.2 | XP_011543170.1 | 16 - 17 | 2188 | 73 | |
GenBank mRNA | AB007864.1 | - | 31 - 32 | 4518 | 73 |
BC008593.1 | - | 2 - 3 | 205 | 73 | |
BC053596.1 | - | 6 - 7 | 905 | 73 | |
BC110650.1 | - | 31 - 32 | 4577 | 73 |