The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080745.1 | NP_001074214.1 | 14 - 15 | 1764 | 82 |
NM_001346.2 | NP_001337.2 | 15 - 16 | 1881 | 82 | |
GenBank mRNA | AB209033.1 | - | 14 - 15 | 1607 | 82 |
AK304440.1 | - | 13 - 14 | 1702 | 82 | |
AK314192.1 | - | 15 - 16 | 1826 | 82 | |
AY737485.1 | - | 13 - 14 | 1459 | 82 | |
BC112363.1 | - | 13 - 14 | 1487 | 82 | |
D26135.1 | - | 15 - 16 | 1860 | 82 |