Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEC16A
Entrez Gene ID
9919
Gene Name
SEC16 homolog A, endoplasmic reticulum export factor
Gene Aliases
KIAA0310, SEC16L, p250
Chromosome Location
Chr.9: 136440096 - 136484552 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276418.1 NP_001263347.1 5 - 6 4248 89
NM_014866.1 NP_055681.1 6 - 7 4238 89
XM_005266134.2 XP_005266191.1 5 - 6 4101 89
XM_011519246.2 XP_011517548.1 6 - 7 4247 89
XM_011519247.1 XP_011517549.1 5 - 6 4104 89
XM_011519248.2 XP_011517550.1 5 - 6 4078 89
XM_011519249.1 XP_011517551.1 5 - 6 4101 89
XM_011519250.2 XP_011517552.1 5 - 6 4101 89
XM_011519251.1 XP_011517553.1 5 - 6 4101 89
XM_011519252.2 XP_011517554.1 5 - 6 4101 89
XM_011519253.1 XP_011517555.1 5 - 6 4101 89
XM_011519254.1 XP_011517556.1 5 - 6 4101 89
XM_011519255.2 XP_011517557.1 5 - 6 4101 89
XM_011519256.1 XP_011517558.1 5 - 6 4101 89
XM_011519257.1 XP_011517559.1 5 - 6 4101 89
XM_011519258.1 XP_011517560.1 5 - 6 4101 89
XM_011519259.1 XP_011517561.1 5 - 6 4101 89
XM_011519260.2 XP_011517562.1 5 - 6 4085 89
XM_011519261.1 XP_011517563.1 5 - 6 4101 89
XM_011519262.1 XP_011517564.1 6 - 7 3390 89
XM_011519263.2 XP_011517565.1 6 - 7 3390 89
XM_011519264.2 XP_011517566.1 5 - 6 4101 89
XM_017015363.1 XP_016870852.1 5 - 6 4101 89
XM_017015364.1 XP_016870853.1 5 - 6 4101 89
GenBank mRNA AB002308.4 - 6 - 7 4238 89
AB593115.1 - 5 - 6 4248 89
BC028183.1 - 1 - 2 105 89
BC125018.1 - 4 - 5 2810 89
BC125019.1 - 5 - 6 2099 89
DQ903855.1 - 4 - 5 3395 89

View Full Product Details