Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SEC16A
- Entrez Gene ID
-
9919
- Gene Name
- SEC16 homolog A, endoplasmic reticulum export factor
- Gene Aliases
-
KIAA0310, SEC16L, p250
- Chromosome Location
-
Chr.9: 136440096 - 136484552 on Build GRCh38
View Full Product Details