Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TPRA1
Entrez Gene ID
131601
Gene Name
transmembrane protein adipocyte associated 1
Gene Aliases
GPR175, TMEM227, TPRA40
Chromosome Location
Chr.3: 127573064 - 127598234 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_073377.1 7 - 8 975 110
XM_017005709.1 XP_016861198.1 8 - 9 1070 110
GenBank mRNA BC017058.1 - 7 - 8 744 110

View Full Product Details