Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNM3
Entrez Gene ID
26052
Gene Name
dynamin 3
Gene Aliases
Dyna III
Chromosome Location
Chr.1: 171841478 - 172418466 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136127.2 NP_001129599.1 14 - 15 1833 68
NM_015569.4 NP_056384.2 14 - 15 1833 68
XM_005245079.1 XP_005245136.1 15 - 16 1846 68
XM_005245080.1 XP_005245137.1 15 - 16 1846 68
XM_017000981.1 XP_016856470.1 15 - 16 1846 68
XM_017000982.1 XP_016856471.1 15 - 16 1846 68
XM_017000983.1 XP_016856472.1 14 - 15 1816 68
XM_017000984.1 XP_016856473.1 14 - 15 1816 68
XM_017000985.1 XP_016856474.1 14 - 15 1816 68
XM_017000986.1 XP_016856475.1 15 - 16 2187 68
XM_017000987.1 XP_016856476.1 15 - 16 1718 68
GenBank mRNA AB020627.2 - 14 - 15 1784 68
AK308992.1 - 14 - 15 1782 68
AL136712.1 - 14 - 15 1811 68

View Full Product Details