Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNM3
Entrez Gene ID
26052
Gene Name
dynamin 3
Gene Aliases
Dyna III
Chromosome Location
Chr.1: 171841478 - 172418466 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136127.2 NP_001129599.1 17 - 18 2220 127
NM_015569.4 NP_056384.2 18 - 19 2232 127
XM_005245079.1 XP_005245136.1 19 - 20 2245 127
XM_005245080.1 XP_005245137.1 18 - 19 2233 127
XM_017000976.1 XP_016856465.1 20 - 21 2389 127
XM_017000977.1 XP_016856466.1 19 - 20 2377 127
XM_017000978.1 XP_016856467.1 19 - 20 2359 127
XM_017000979.1 XP_016856468.1 20 - 21 2389 127
XM_017000980.1 XP_016856469.1 20 - 21 2107 127
XM_017000981.1 XP_016856470.1 18 - 19 2233 127
XM_017000982.1 XP_016856471.1 18 - 19 2233 127
XM_017000983.1 XP_016856472.1 18 - 19 2215 127
XM_017000984.1 XP_016856473.1 17 - 18 2203 127
XM_017000985.1 XP_016856474.1 17 - 18 2203 127
XM_017000986.1 XP_016856475.1 18 - 19 2574 127
XM_017000987.1 XP_016856476.1 19 - 20 2117 127
GenBank mRNA AB020627.2 - 17 - 18 2171 127
AL035110.1 - 1 - 2 160 127
AL136712.1 - 18 - 19 2210 127

View Full Product Details