Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LARP1B
Entrez Gene ID
55132
Gene Name
La ribonucleoprotein domain family member 1B
Gene Aliases
LARP2
Chromosome Location
Chr.4: 128060655 - 128222931 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018078.3 NP_060548.2 17 - 18 2518 152
XM_011532057.2 XP_011530359.2 19 - 20 3404 152
XM_011532058.2 XP_011530360.2 17 - 18 3331 152
XM_011532059.2 XP_011530361.2 19 - 20 3323 152
XM_011532060.2 XP_011530362.2 16 - 17 3283 152
XM_011532061.2 XP_011530363.1 18 - 19 3265 152
XM_011532062.2 XP_011530364.2 17 - 18 3250 152
XM_011532063.2 XP_011530365.2 17 - 18 3238 152
XM_011532064.2 XP_011530366.2 16 - 17 3202 152
XM_011532065.2 XP_011530367.1 15 - 16 3144 152
XM_011532067.1 XP_011530369.1 19 - 20 2602 152
XM_011532068.1 XP_011530370.1 18 - 19 2435 152
XM_011532069.1 XP_011530371.1 15 - 16 2315 152
XM_011532075.1 XP_011530377.1 17 - 18 2272 152
XM_011532076.2 XP_011530378.1 17 - 18 2602 152
XM_011532077.2 XP_011530379.1 13 - 14 1812 152
XM_017008338.1 XP_016863827.1 19 - 20 3404 152
XM_017008339.1 XP_016863828.1 17 - 18 3319 152
XM_017008340.1 XP_016863829.1 19 - 20 3037 152
XM_017008341.1 XP_016863830.1 19 - 20 2550 152
XM_017008342.1 XP_016863831.1 17 - 18 2408 152
XM_017008343.1 XP_016863832.1 16 - 17 2482 152
XM_017008344.1 XP_016863833.1 16 - 17 2917 152
XM_017008346.1 XP_016863835.1 17 - 18 2363 152
XM_017008349.1 XP_016863838.1 17 - 18 2530 152
XM_017008350.1 XP_016863839.1 14 - 15 1940 152
XM_017008351.1 XP_016863840.1 14 - 15 2082 152
GenBank mRNA AK027164.1 - 4 - 5 486 152
AL137759.1 - 14 - 15 2252 152
BC030516.1 - 3 - 4 501 152
BC062606.1 - 3 - 4 468 152
BX537937.1 - 5 - 6 693 152

View Full Product Details