Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ART3
Entrez Gene ID
419
Gene Name
ADP-ribosyltransferase 3
Gene Aliases
ARTC3
Chromosome Location
Chr.4: 76011180 - 76112802 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130016.2 NP_001123488.1 10 - 11 1138 93
XM_017008209.1 XP_016863698.1 8 - 9 983 93
GenBank mRNA AY550342.1 - 8 - 9 260 93
AY550343.1 - 7 - 8 227 93
BC008397.1 - 10 - 11 1125 93
BT007249.1 - 9 - 10 1004 93
CR541664.1 - 9 - 10 1004 93

View Full Product Details