Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CISD2
Entrez Gene ID
493856
Gene Name
CDGSH iron sulfur domain 2
Gene Aliases
ERIS, Miner1, NAF-1, WFS2, ZCD2
Chromosome Location
Chr.4: 102868978 - 102892807 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008388.4 NP_001008389.1 2 - 3 424 92
GenBank mRNA AK292134.1 - 2 - 3 361 92
BC032300.1 - 2 - 3 361 92
BX537971.1 - 2 - 3 362 92
DN993861.1 - 2 - 3 429 92

View Full Product Details