The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178527.3 | NP_848622.2 | 18 - 19 | 2738 | 86 |
XM_011509426.1 | XP_011507728.1 | 18 - 19 | 2738 | 86 | |
XM_017001065.1 | XP_016856554.1 | 18 - 19 | 2433 | 86 | |
XM_017001066.1 | XP_016856555.1 | 18 - 19 | 2427 | 86 | |
XM_017001067.1 | XP_016856556.1 | 17 - 18 | 2618 | 86 | |
XM_017001068.1 | XP_016856557.1 | 17 - 18 | 2637 | 86 | |
XM_017001069.1 | XP_016856558.1 | 17 - 18 | 2332 | 86 | |
XM_017001070.1 | XP_016856559.1 | 17 - 18 | 2326 | 86 | |
XM_017001071.1 | XP_016856560.1 | 18 - 19 | 2345 | 86 | |
XM_017001072.1 | XP_016856561.1 | 16 - 17 | 2471 | 86 | |
XM_017001073.1 | XP_016856562.1 | 17 - 18 | 2244 | 86 | |
XM_017001074.1 | XP_016856563.1 | 16 - 17 | 2173 | 86 | |
XM_017001075.1 | XP_016856564.1 | 11 - 12 | 1500 | 86 | |
XM_017001076.1 | XP_016856565.1 | 8 - 9 | 1107 | 86 | |
GenBank mRNA | AK128104.1 | - | 18 - 19 | 2738 | 86 |
AK302731.1 | - | 15 - 16 | 2135 | 86 | |
BC042592.1 | - | 18 - 19 | 2685 | 86 | |
BX648119.1 | - | 11 - 12 | 1649 | 86 |