The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178527.3 | NP_848622.2 | 20 - 21 | 2931 | 109 |
XM_011509426.1 | XP_011507728.1 | 20 - 21 | 2931 | 109 | |
XM_017001065.1 | XP_016856554.1 | 20 - 21 | 2626 | 109 | |
XM_017001066.1 | XP_016856555.1 | 20 - 21 | 2620 | 109 | |
XM_017001067.1 | XP_016856556.1 | 19 - 20 | 2811 | 109 | |
XM_017001068.1 | XP_016856557.1 | 19 - 20 | 2830 | 109 | |
XM_017001069.1 | XP_016856558.1 | 19 - 20 | 2525 | 109 | |
XM_017001070.1 | XP_016856559.1 | 19 - 20 | 2519 | 109 | |
XM_017001071.1 | XP_016856560.1 | 20 - 21 | 2538 | 109 | |
XM_017001072.1 | XP_016856561.1 | 18 - 19 | 2664 | 109 | |
XM_017001073.1 | XP_016856562.1 | 19 - 20 | 2437 | 109 | |
XM_017001074.1 | XP_016856563.1 | 18 - 19 | 2366 | 109 | |
XM_017001075.1 | XP_016856564.1 | 13 - 14 | 1693 | 109 | |
XM_017001076.1 | XP_016856565.1 | 10 - 11 | 1300 | 109 | |
GenBank mRNA | AK128104.1 | - | 20 - 21 | 2931 | 109 |
BC042592.1 | - | 20 - 21 | 2878 | 109 | |
BX648119.1 | - | 13 - 14 | 1842 | 109 |