The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127698.1 | NP_001121170.1 | 24 - 25 | 2389 | 122 |
NM_001127699.1 | NP_001121171.1 | 24 - 25 | 2389 | 122 | |
NM_006846.3 | NP_006837.2 | 24 - 25 | 2389 | 122 | |
XM_011537551.2 | XP_011535853.1 | 24 - 25 | 2475 | 122 | |
GenBank mRNA | AJ228139.2 | - | 24 - 25 | 2359 | 122 |
AK025197.1 | - | 3 - 4 | 298 | 122 | |
AK299455.1 | - | 24 - 25 | 2324 | 122 | |
AK301660.1 | - | 24 - 25 | 2322 | 122 | |
DQ149927.1 | - | 24 - 25 | 2389 | 122 | |
DQ149928.1 | - | 24 - 25 | 2389 | 122 | |
DQ149929.1 | - | 24 - 25 | 2389 | 122 |