Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPTBN5
Entrez Gene ID
51332
Gene Name
spectrin beta, non-erythrocytic 5
Gene Aliases
BSPECV, HUBSPECV, HUSPECV
Chromosome Location
Chr.15: 41848146 - 41894077 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016642.3 NP_057726.4 14 - 15 3035 104
XM_017022299.1 XP_016877788.1 13 - 14 2893 104
XM_017022300.1 XP_016877789.1 13 - 14 2893 104
XM_017022301.1 XP_016877790.1 9 - 10 2344 104
XM_017022303.1 XP_016877792.1 13 - 14 2893 104
XM_017022304.1 XP_016877793.1 13 - 14 2893 104
XM_017022305.1 XP_016877794.1 13 - 14 2893 104
GenBank mRNA AF233523.1 - 14 - 15 3035 104

View Full Product Details