Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPTBN5
Entrez Gene ID
51332
Gene Name
spectrin beta, non-erythrocytic 5
Gene Aliases
BSPECV, HUBSPECV, HUSPECV
Chromosome Location
Chr.15: 41848146 - 41894077 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016642.3 NP_057726.4 39 - 40 7144 67
XM_017022299.1 XP_016877788.1 38 - 39 7002 67
XM_017022300.1 XP_016877789.1 38 - 39 7002 67
XM_017022301.1 XP_016877790.1 34 - 35 6453 67
XM_017022302.1 XP_016877791.1 26 - 27 4251 67
XM_017022303.1 XP_016877792.1 38 - 39 7002 67
GenBank mRNA AF233523.1 - 39 - 40 7144 67

View Full Product Details