Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPTBN5
Entrez Gene ID
51332
Gene Name
spectrin beta, non-erythrocytic 5
Gene Aliases
BSPECV, HUBSPECV, HUSPECV
Chromosome Location
Chr.15: 41848146 - 41894077 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016642.3 NP_057726.4 66 - 67 11152 95
XM_017022299.1 XP_016877788.1 64 - 65 11130 95
XM_017022300.1 XP_016877789.1 65 - 66 11010 95
XM_017022301.1 XP_016877790.1 60 - 61 10581 95
XM_017022302.1 XP_016877791.1 52 - 53 8379 95
GenBank mRNA AF233523.1 - 66 - 67 11152 95

View Full Product Details