The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014568.1 | NP_055383.1 | 4 - 5 | 2134 | 92 |
XM_017003237.1 | XP_016858726.1 | 4 - 5 | 2414 | 92 | |
GenBank mRNA | AF154107.1 | - | 4 - 5 | 890 | 92 |
AJ245539.1 | - | 4 - 5 | 1063 | 92 | |
AK292154.1 | - | 4 - 5 | 2095 | 92 | |
AY277591.1 | - | 4 - 5 | 2134 | 92 | |
BC142676.1 | - | 4 - 5 | 2364 | 92 | |
BC144211.1 | - | 5 - 6 | 2077 | 92 |