The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014568.1 | NP_055383.1 | 7 - 8 | 2694 | 105 |
XM_017003237.1 | XP_016858726.1 | 7 - 8 | 2974 | 105 | |
GenBank mRNA | AF154107.1 | - | 7 - 8 | 1450 | 105 |
AJ245539.1 | - | 7 - 8 | 1623 | 105 | |
AJ505956.1 | - | 1 - 2 | 126 | 105 | |
AK292154.1 | - | 7 - 8 | 2655 | 105 | |
AY277591.1 | - | 7 - 8 | 2694 | 105 | |
BC142676.1 | - | 7 - 8 | 2924 | 105 | |
BC144211.1 | - | 8 - 9 | 2637 | 105 |