Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYOT
Entrez Gene ID
9499
Gene Name
myotilin
Gene Aliases
LGMD1, LGMD1A, MFM3, TTID, TTOD
Chromosome Location
Chr.5: 137867282 - 137887851 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135940.1 NP_001129412.1 6 - 7 639 147
NM_001300911.1 NP_001287840.1 7 - 8 770 147
NM_006790.2 NP_006781.1 6 - 7 1121 147
XM_017010060.1 XP_016865549.1 7 - 8 1339 147
XM_017010061.1 XP_016865550.1 5 - 6 772 147
XM_017010062.1 XP_016865551.1 6 - 7 1208 147
GenBank mRNA AF133820.2 - 6 - 7 1106 147
AF144477.1 - 6 - 7 1092 147
AK300076.1 - 7 - 8 750 147
AK300088.1 - 6 - 7 637 147
AK307620.1 - 7 - 8 984 147
BC005376.1 - 6 - 7 1101 147
KU178476.1 - 5 - 6 812 147

View Full Product Details