Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ECI1
Entrez Gene ID
1632
Gene Name
enoyl-CoA delta isomerase 1
Gene Aliases
DCI
Chromosome Location
Chr.16: 2239872 - 2251601 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001919.3 NP_001910.2 5 - 6 600 64
GenBank mRNA AK291127.1 - 5 - 6 600 64
BC000762.2 - 5 - 6 565 64
BC009631.1 - 6 - 7 542 64
BC019316.1 - 5 - 6 565 64
BQ773923.1 - 2 - 3 64
L24774.1 - 4 - 5 457 64
Z25820.1 - 5 - 6 573 64

View Full Product Details