The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261816.1 | NP_001248745.1 | 7 - 8 | 928 | 57 |
NM_002613.4 | NP_002604.1 | 7 - 8 | 928 | 57 | |
NM_031268.5 | NP_112558.2 | 4 - 5 | 547 | 57 | |
XM_011522521.2 | XP_011520823.1 | 7 - 8 | 4616 | 57 | |
XM_011522523.2 | XP_011520825.1 | 4 - 5 | 4240 | 57 | |
XM_017023271.1 | XP_016878760.1 | 7 - 8 | 4621 | 57 | |
XM_017023272.1 | XP_016878761.1 | 7 - 8 | 1177 | 57 | |
XM_017023273.1 | XP_016878762.1 | 7 - 8 | 886 | 57 | |
XM_017023274.1 | XP_016878763.1 | 8 - 9 | 1024 | 57 | |
XM_017023275.1 | XP_016878764.1 | 4 - 5 | 4246 | 57 | |
XM_017023276.1 | XP_016878765.1 | 4 - 5 | 490 | 57 | |
XM_017023277.1 | XP_016878766.1 | 5 - 6 | 588 | 57 | |
GenBank mRNA | AB209835.1 | - | 7 - 8 | 895 | 57 |
AF017995.1 | - | 7 - 8 | 859 | 57 | |
AJ785968.1 | - | 6 - 7 | 1562 | 57 | |
AK131060.1 | - | 7 - 8 | 862 | 57 | |
AK222581.1 | - | 7 - 8 | 871 | 57 | |
BC006339.2 | - | 4 - 5 | 508 | 57 | |
BC012103.1 | - | 7 - 8 | 888 | 57 | |
BC033494.1 | - | 4 - 5 | 460 | 57 | |
CR536517.1 | - | 7 - 8 | 779 | 57 | |
Y15056.1 | - | 7 - 8 | 858 | 57 |