Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATF6
Entrez Gene ID
22926
Gene Name
activating transcription factor 6
Gene Aliases
ACHM7, ATF6A
Chromosome Location
Chr.1: 161766244 - 161964070 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007348.3 NP_031374.2 14 - 15 1838 144
XM_006711224.1 XP_006711287.1 14 - 15 1759 144
XM_011509308.1 XP_011507610.1 14 - 15 1819 144
XM_011509309.1 XP_011507611.1 14 - 15 1816 144
GenBank mRNA AB015856.1 - 14 - 15 1789 144
AF005887.1 - 14 - 15 1763 144
AK290498.1 - 14 - 15 1762 144

View Full Product Details