The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001248.3 | NP_001239.2 | 5 - 6 | 561 | 142 |
NM_001291960.1 | NP_001278889.1 | 5 - 6 | 602 | 142 | |
NM_001291961.1 | NP_001278890.1 | 5 - 6 | 561 | 142 | |
XM_011534266.2 | XP_011532568.1 | 5 - 6 | 561 | 142 | |
GenBank mRNA | AF034840.2 | - | 5 - 6 | 522 | 142 |
AF039917.1 | - | 5 - 6 | 525 | 142 | |
AK313322.1 | - | 5 - 6 | 587 | 142 | |
BC029869.1 | - | 4 - 5 | 465 | 142 | |
BX952692.1 | - | 5 - 6 | 536 | 142 |