Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ENAM
Entrez Gene ID
10117
Gene Name
enamelin
Gene Aliases
ADAI, AI1C, AIH2
Chromosome Location
Chr.4: 70627471 - 70646819 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031889.2 NP_114095.2 6 - 7 758 81
XM_006714056.3 XP_006714119.1 5 - 6 2507 81
GenBank mRNA AF125373.1 - 6 - 7 758 81
BC117308.1 - 5 - 6 547 81
BC117310.1 - 5 - 6 547 81

View Full Product Details