The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007332.2 | NP_015628.2 | 15 - 16 | 2081 | 149 |
XM_011517624.2 | XP_011515926.1 | 16 - 17 | 6897 | 149 | |
XM_011517625.2 | XP_011515927.1 | 17 - 18 | 7056 | 149 | |
XM_017013946.1 | XP_016869435.1 | 16 - 17 | 3359 | 149 | |
GenBank mRNA | AK307942.1 | - | 15 - 16 | 2096 | 149 |
Y10601.1 | - | 15 - 16 | 2080 | 149 |