The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007332.2 | NP_015628.2 | 6 - 7 | 976 | 62 |
XM_011517624.2 | XP_011515926.1 | 7 - 8 | 5792 | 62 | |
XM_011517625.2 | XP_011515927.1 | 8 - 9 | 5951 | 62 | |
XM_017013946.1 | XP_016869435.1 | 7 - 8 | 2254 | 62 | |
GenBank mRNA | AK307942.1 | - | 6 - 7 | 991 | 62 |
Y10601.1 | - | 6 - 7 | 975 | 62 |