The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007231.4 | NP_009162.1 | 11 - 12 | 1638 | 138 |
GenBank mRNA | AF151978.1 | - | 11 - 12 | 1595 | 138 |
AK313390.1 | - | 11 - 12 | 138 | ||
BC093710.1 | - | 11 - 12 | 1529 | 138 | |
BC093712.1 | - | 11 - 12 | 1529 | 138 |