The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253837.1 | NP_001240766.1 | 13 - 14 | 2529 | 61 |
NM_016112.2 | NP_057196.2 | 13 - 14 | 2507 | 61 | |
XM_011540323.2 | XP_011538625.1 | 14 - 15 | 2070 | 61 | |
XM_011540325.2 | XP_011538627.1 | 15 - 16 | 2180 | 61 | |
XM_017016875.1 | XP_016872364.1 | 14 - 15 | 2018 | 61 | |
XM_017016876.1 | XP_016872365.1 | 12 - 13 | 1776 | 61 | |
GenBank mRNA | AF053316.1 | - | 13 - 14 | 1999 | 61 |
AF073481.1 | - | 13 - 14 | 2507 | 61 | |
AF092170.1 | - | 11 - 12 | 1786 | 61 | |
AF094827.1 | - | 12 - 13 | 1839 | 61 | |
BC025665.1 | - | 13 - 14 | 2204 | 61 | |
DQ084244.1 | - | 13 - 14 | 1983 | 61 |