The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253837.1 | NP_001240766.1 | 2 - 3 | 749 | 80 |
NM_016112.2 | NP_057196.2 | 2 - 3 | 727 | 80 | |
GenBank mRNA | AF053316.1 | - | 2 - 3 | 219 | 80 |
AF073481.1 | - | 2 - 3 | 727 | 80 | |
AF094827.1 | - | 1 - 2 | 59 | 80 | |
BC025665.1 | - | 2 - 3 | 424 | 80 |