The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253837.1 | NP_001240766.1 | 7 - 8 | 1756 | 74 |
NM_016112.2 | NP_057196.2 | 7 - 8 | 1734 | 74 | |
XM_011540323.2 | XP_011538625.1 | 8 - 9 | 1297 | 74 | |
XM_011540325.2 | XP_011538627.1 | 9 - 10 | 1407 | 74 | |
XM_017016875.1 | XP_016872364.1 | 8 - 9 | 1245 | 74 | |
XM_017016876.1 | XP_016872365.1 | 6 - 7 | 1003 | 74 | |
GenBank mRNA | AF053316.1 | - | 7 - 8 | 1226 | 74 |
AF073481.1 | - | 7 - 8 | 1734 | 74 | |
AF094827.1 | - | 6 - 7 | 1066 | 74 | |
BC025665.1 | - | 7 - 8 | 1431 | 74 | |
DQ084244.1 | - | 7 - 8 | 1210 | 74 |