The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002013.3 | NP_002004.1 | 4 - 5 | 520 | 103 |
XM_011536565.2 | XP_011534867.1 | 4 - 5 | 2541 | 103 | |
XM_017021092.1 | XP_016876581.1 | 4 - 5 | 469 | 103 | |
XM_017021093.1 | XP_016876582.1 | 4 - 5 | 2541 | 103 | |
GenBank mRNA | AK222993.1 | - | 4 - 5 | 469 | 103 |
AK311915.1 | - | 4 - 5 | 458 | 103 | |
BC016288.1 | - | 5 - 6 | 858 | 103 | |
BC020809.1 | - | 4 - 5 | 458 | 103 | |
BI598048.1 | - | 4 - 5 | 525 | 103 | |
BT006904.1 | - | 4 - 5 | 447 | 103 | |
M96256.1 | - | 4 - 5 | 470 | 103 |