The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002013.3 | NP_002004.1 | 6 - 7 | 695 | 116 |
XM_011536565.2 | XP_011534867.1 | 6 - 7 | 2716 | 116 | |
GenBank mRNA | AK222993.1 | - | 6 - 7 | 644 | 116 |
BC016288.1 | - | 7 - 8 | 1033 | 116 | |
BC020809.1 | - | 6 - 7 | 633 | 116 |