The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003566.3 | NP_003557.2 | 20 - 21 | 3040 | 108 |
XM_011538814.2 | XP_011537116.1 | 21 - 22 | 3146 | 108 | |
XM_011538815.2 | XP_011537117.1 | 20 - 21 | 2857 | 108 | |
XM_017020018.1 | XP_016875507.1 | 20 - 21 | 2808 | 108 | |
GenBank mRNA | L40157.1 | - | 20 - 21 | 2911 | 108 |
X78998.1 | - | 20 - 21 | 2846 | 108 |