The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271028.1 | NP_001257957.1 | 8 - 9 | 1629 | 85 |
NM_021153.3 | NP_066976.1 | 8 - 9 | 1629 | 85 | |
NR_073130.1 | 8 - 9 | 1629 | 85 | ||
XM_011525931.2 | XP_011524233.1 | 8 - 9 | 1596 | 85 | |
XM_017025711.1 | XP_016881200.1 | 8 - 9 | 1518 | 85 | |
GenBank mRNA | AJ007607.1 | - | 8 - 9 | 1470 | 85 |
AK304000.1 | - | 8 - 9 | 1579 | 85 | |
AK315257.1 | - | 8 - 9 | 1631 | 85 | |
AY358654.1 | - | 7 - 8 | 1368 | 85 |