Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRA1
Entrez Gene ID
10011
Gene Name
steroid receptor RNA activator 1
Gene Aliases
SRA, SRAP, STRAA1, pp7684
Chromosome Location
Chr.5: 140550067 - 140558093 on Build GRCh38
Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035235.3 NP_001030312.2
1 - 2 664 58
GenBank mRNA AF293024.1 - LIT1246
SRA
1 - 2 248 58
AF293025.1 - LIT1250
SRA
1 - 2 248 58
AF293026.1 - LIT1251
SRA
1 - 2 248 58
AF318361.1 -
1 - 2 665 58
AK054960.1 -
1 - 2 881 58
CX165918.1 -
1 - 2 286 58

View Full Product Details