The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004739.3 | NP_004730.2 | 16 - 17 | 2082 | 69 |
XM_017018561.1 | XP_016874050.1 | 16 - 17 | 1934 | 69 | |
GenBank mRNA | AB016591.1 | - | 16 - 17 | 2015 | 69 |
AF295807.1 | - | 16 - 17 | 1692 | 69 | |
AK301569.1 | - | 16 - 17 | 1917 | 69 | |
BC023656.2 | - | 16 - 17 | 1878 | 69 | |
BC053650.1 | - | 16 - 17 | 1900 | 69 | |
CR749481.1 | - | 14 - 15 | 1603 | 69 |